Peqlab HX-1 / HX-2, Blocks for Block Heaters, Digital
Supplier: VWR Collection
PEQL91-D1102AEA 174 EUR
PEQL91-D1102A PEQL91-D1105 PEQL91-D1105A PEQL91-D1113A PEQL91-D1117A PEQL91-D1150A PEQL91-D1500A
Peqlab HX-1 / HX-2, Blocks for Block Heaters, Digital
Block Heaters Blocks
Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy.
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Thermometer wells available in all blocks except those used with plates.