Chemical resistant coverall, BioSafe, Type 4B, SAFE®
Supplier: DACH SCHUTZBEKLEIDUNG
561/L 561/M 561/XL 561/XXL 561/XXXL
DACH561/LEA 0 EUR
DACH561/L DACH561/M DACH561/XL DACH561/XXL DACH561/XXXL
Chemical resistant coverall, BioSafe, Type 4B, SAFE®
Disposable Garments
The BioSafe coverall protects the user against chemical and biological hazards, while at the same time ensuring complete freedom of mobility.
  • Spray-tight material
  • Knitted cuffs with thumb loops
  • Sealed and taped seams
  • Antistatic material
The multi-layered composite material with sealed and taped seams ensures mechanical stability and tear resistance. Due to its unique features, the BioSafe coverall is used predominantly in the medical and chemical field.
Certifikace: Meets the requirements for a chemical protective coverall Type 4B, 5B, 6B. Also complies with the European standards EN 14126, EN 1073-2 and 1149-5.