Trifluoroacetic acid 99.01-101.0% (by titrimetric analysis), GPR RECTAPUR®
Supplier: VWR Chemicals
Danger
Synonyms: TFA
85903.150EA 215 EUR
85903.150 85903.180 85903.290
Trifluoroacetic acid 99.01-101.0% (by titrimetric analysis), GPR RECTAPUR®
Trifluoroacetic acid
Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8
Melting Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (Titration) 99.0-101.0 %w/w
Relative density (d 20°C/4°C) 1.487-1.490
Identity (IR) Complying
Water (KF) max. 0.1 %w/w