Polypropylene glycol, M.W. 1200
Supplier: Thermo Scientific
Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG
47361.30 47361.18
47361.30EA 65.7 EUR
47361.30 47361.18
Polypropylene glycol, M.W. 1200
Polypropylene glycol
Formula: H(OCH₂CH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8