Pyridine, anhydrous ≥99.5%, AcroSeal®
Supplier: Thermo Scientific
|
Danger
|
Packaged under argon.
47323.AEEA
70.4
EUR
47323.AE
47323.K2
Pyridine, anhydrous ≥99.5%, AcroSeal®
Pyridine
AcroSeal™ septum bottled solvent
- Supports optimum performance of your moisture-sensitive work
Formula:
C₅H₅N MW: 79,1 g/mol Boiling Pt: 115 °C Melting Pt: –42 °C Density: 0,978 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3, Merck Index: 14,7970 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...