VWR®, Refractive index standard kits for calibration according to Ph. Eur. 2.2.6
Supplier: VWR Chemicals
85757.001EA
398
EUR
85757.001
85758.001
VWR®, Refractive index standard kits for calibration according to Ph. Eur. 2.2.6
Standards
Refractive Index Standards
These standards are for the calibration of refractometers prior to use.