Tyramide amplification kit with HRP goat, anti-rabbit and CF® dye or biotin tyramide
Supplier: Biotium
33001 33004 33007 33010 33013 33016 33019
33001.EA 842 EUR
33001. 33004. 33007. 33010. 33013. 33016. 33019.
Tyramide amplification kit with HRP goat, anti-rabbit and CF® dye or biotin tyramide
Protein Linking and Labeling Systems
Kits for increasing immunofluorescence sensitivity using HRP-catalysed tyramide signal amplification with the choice of next-generation fluorescent CF® dyes or biotin.
  • Increase sensitivity by up to 100-fold
  • Use lower primary antibody concentrations
  • Boost signal in challenging samples, like human FFPE tissues
  • Choose from six bright and photostable CF® dyes or biotin
Tyramide amplification, sometimes called Catalysed Reporter Deposition (CARD), is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labelling of a target protein or nucleic acid sequence in situ. The tyramide amplification method has been reported to increase the sensitivity by up to 100-fold compared to conventional avidin-biotin detection. Multiple rounds of tyramide signal amplification can be performed for multiplex detection.
The HRP goat anti-rabbit antibody provided in the kits has been adsorbed against human, mouse, and rat serum to minimise cross-reactivity. The kits include choice of six CF® dye tyramides or biotin tyramide. CF® dyes are line of next-generation fluorescent dyes, with advantages in brightness and photostability compared to other dyes.
Informace pro objednávání: Each kit contains reagents for staining 50 to 150 slides; the actual number of samples that can be stained will depend on staining volumes used. Tyramide amplification buffer, CF® dye tyramides, and other tyramide compounds are also available separately. Note: Endogenous biotin blocking reagents (not provided) may be required for streptavidin/biotin detection in cells or tissues.
Delivery information: Kit components: CF® dye or biotin-tyramide stock solution, HRP goat anti-rabbit IgG (highly cross-adsorbed), BSA for blocking, Tyramide amplification buffer plus, 30% Hydrogen peroxide.
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Learn more