Counting chambers, Nageotte
Supplier: Hecht
Nageotte
631-0928EA 50.2 EUR
631-0928 630-2741
Counting chambers, Nageotte
Slides Counting Chambers
These counting chambers are used for special applications in lumbar fluid.
  • Subdivision: 40 rectangles of 10×0,25 mm each
  • Centre of ruling marked by double lines