VWR® Joseph Paper, Grade 551
Supplier: VWR Collection
111-5005EA 52 EUR
111-5005 111-5007 111-5006 111-5009 111-2495
VWR® Joseph Paper, Grade 551
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
Absorbent paper, suitable for cleaning and drying glass containers, tubes, microscopic plates, bottles, and more.
Thickness: 0,07 mm
Weight: 25 g/m²