VWR® RC-10 Recirculating Chillers
Supplier: VWR Collection
462-0137EA 2580 EUR
462-0137 462-0138 462-0139
VWR® RC-10 Recirculating Chillers
Circulating Thermostats Closed Circulator Systems Cooling Circulators
VWR® recirculating chillers are ideal for use with rotary evaporators, as they ensure constant, reproducible cooling conditions and avoid wasting tap water thereby increasing cost savings. Designed to be user friendly, the units have an easily accessible filling port on top of the unit and a drain tap at the back. The chillers have a stainless steel reservoir, operate using R134 refrigerant and can be used in the permitted ambient temperature range of 15 to 32 °C.
  • Temperature set point can be adjusted by using the keys on the command panel (except the basic model which has a fixed set point at 7 °C)
  • The Duo chiller can cool two cooling cycles directly connected on the double outlets and inlets
  • Regulate the temperature of the cooling cycle between −10 and +40 °C (use water and glycol mixture (70:30 mixture) for temperatures below +5 °C)
  • Indicator on front panel for perfect view of the liquid level
Max. flow rate (with no counter pressure) / max. pressure (at zero flow)
RC-10 Basic: 14 L/min, 0,4 bar
RC-10 Digital: 8 L/min, 0,6 bar
RC-10 Duo: 11 L/min, 1,0 bar
Delivery information: Basic and Digital units supplied with 2×8 mm Ø hose connections, 2× screw nuts M 16×1, 2× PVC hose (1500 mm) and 2× hose clamps, Duo models have 4 of each of these. Includes EU, UK and CH power cables.
Upozornení: Note: Cooling capacity at +15 °C
Fixed