VWR® ARISTAR®, Multi-Element Quality Control Standard 1 in dil. Nitric Acid for ICP-MS
Supplier: Avantor
VWR® ARISTAR®
456592QEA 319 EUR
456592Q
VWR® ARISTAR®, Multi-Element Quality Control Standard 1 in dil. Nitric Acid for ICP-MS
Standards Cationic Standards Multi-Element Standards
9 Elements in 2% nitric acid solution.
9 Elements 10 mg/L : Be, Bi, Ce, Co, In, Pb, Mg, Ni, U
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Certifikace: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022