Dipotassium-L(+)-tartrate hemihydrate ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals

27048.291EA 129 EUR
27048.291
Dipotassium-L(+)-tartrate hemihydrate ≥98%, GPR RECTAPUR®
Dipotassium-L(+)-tartrate hemihydrate
Formula: KOOC(CHOH)₂COOK·0.5H₂O
MW: 235,28 g/mol
Density: 1,943 g/cm³ (20 °C)
MDL Number: MFCD00150136
CAS Number: 6100-19-2
Merck Index: 12,07849

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 50 ppm
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 0.02 %
Fe (Iron) Max. 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR