Fluorescein-12-dUTP, lyophilized powder
Supplier: Biotium
40063.EA
517
EUR
40063.
Fluorescein-12-dUTP, lyophilized powder
Fluorescein-12-dUTP
Fluorescein-12-dUTP can be used to synthesize fluorecently labeled DNA probes for in-situ hybridization, microarray or blotting techniques.
Fluorescein has absorption/emission at 494/521 nm.
Upozornení: Store Fluorescein-12-dUTP at –20 °C or –70 °C, protected from light.