VWR® 210 Heating Recirculators
Supplier: VWR
VWR®
473-0031EA 2470 EUR
473-0031 473-0030 473-0029
VWR® 210 Heating Recirculators
Circulating Thermostats Closed Circulator Systems Heating Circulators
Small, low cost recirculator that handles applications such as thawing plasma and tempering photographic solutions. The sealed reservoir can control closed loop systems only.
Features stainless steel, PVC, brass and nylon wetted parts and ½" NPT pump inlet/outlet.
Low liquid and over-temperature cut-offs.
Max. flowrate: 11 L/min.
Upozornení: * RT = Ambient.