VWR® HPLC Microsyringes, Rheodyne
Supplier: VWR Collection
549-0534EA 59 EUR
549-0534 549-0535
VWR® HPLC Microsyringes, Rheodyne
Syringes Syringes for Chromatography
Fine fixed needle, with special polished blunt tip, specific length and diameter, ensures a complete seal, preventing damage to needle seals and rotor.
549-0534: Super-flexible plunger.
Length in ordering table refers to needle length.