Gold ≥99.999% (metals basis), foil, Premion®, Thickness: 0.5 mm (0.02 in)
Supplier: Thermo Fisher Scientific
44164.FF 44164.FI
44164.FFEA 0 EUR
44164.FF 44164.FI
Gold ≥99.999% (metals basis), foil, Premion®, Thickness: 0.5 mm (0.02 in)
Gold
Purity: 99.999% Physical Form: ≈6g/25x25mm Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526