Methanol-[D₄], Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol
MQ200
439029-10G 439029-50G
SIAL439029-10GEA 199 EUR
SIAL439029-10G SIAL439029-50G
Methanol-[D₄], Sigma-Aldrich®
Methanol-[D4]
Formula: D₃COD
MW: 36.01 g/mol
Boiling Pt: 65.3
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3 II