Toluene-[D₈], Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIAL269875-25GEA
262
EUR
SIAL269875-25G
Toluene-[D₈], Sigma-Aldrich®
Toluene-[D₈]
Formula:
C₆D₅CD₃ MW: 100,08 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –85 °C Density: 0,94 g/cm³ (20 °C) Flash Pt: 4 °C (closed cup) |
MDL Number:
MFCD00044638 CAS Number: 2037-26-5 UN: 1294 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...