Acetonitrile-[D₃], Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
SIAL699543-50GEA 389 EUR
SIAL699543-50G SIAL699543-10G
Acetonitrile-[D₃], Sigma-Aldrich®
Acetonitrile-[D₃]
Formula: D₃CCN
MW: 44,03 g/mol
Boiling Pt: 80,7 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0,844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
MDL Number: MFCD00001881
CAS Number: 2206-26-0
UN: 1648
ADR: 3,II