L(+)-Rhamnose monohydrate
Supplier: PanReac AppliChem
Synonyms: (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate
A4336.0010 A4336.0100
A4336.0010EA 103 EUR
A4336.0010 A4336.0100
L(+)-Rhamnose monohydrate
L(+)-Rhamnose monohydrate
Formula: C₆H₁₂O₅·1H₂O
MW: 182.17 g/mol
Melting Pt: 85…95 °C
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149363
CAS Number: 10030-85-0
EINECS: 222-793-4
Merck Index: 12,08338