L(+)-Rhamnose monohydrate
Supplier: Roth Carl
Synonyms:
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate
4655.1
4655.2
ROTH4655.1EA
0
EUR
ROTH4655.1
ROTH4655.2
L(+)-Rhamnose monohydrate
L(+)-Rhamnose monohydrate
Formula:
C₆H₁₂O₅·1H₂O MW: 182.17 g/mol Melting Pt: 85…95 °C Density: 1.47 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149363 CAS Number: 10030-85-0 EINECS: 222-793-4 Merck Index: 12,08338 |