L(+)-Rhamnose monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal hydrate, 6-Deoxy-L(+)-mannose monohydrate
MQ200
R3875-100G R3875-10G R3875-10MG R3875-25G R3875-50G R3875-5G
SIALR3875-100GEA 596 EUR
SIALR3875-100G SIALR3875-10G SIALR3875-10MG SIALR3875-25G SIALR3875-50G SIALR3875-5G
L(+)-Rhamnose monohydrate, Sigma-Aldrich®
L(+)-Rhamnose monohydrate
Formula: C₆H₁₂O₅·1H₂O
MW: 182.17 g/mol
Melting Pt: 85…95 °C
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149363
CAS Number: 10030-85-0
EINECS: 222-793-4
Merck Index: 12,08338