Hexacarbonylmolybdenum, Sigma-Aldrich®

Supplier: Merck

Synonyms: Molybdenum carbonyl, Hexacarbonylmolybdenum(0), Molybdenumhexacarbonyl, Molybdenum hexacarbonyl

M-Clarity™ quality level = MQ100

199958-10G 199958-250G
SIAL199958-10GEA 101 EUR
SIAL199958-10G SIAL199958-250G
Hexacarbonylmolybdenum, Sigma-Aldrich®
Hexacarbonylmolybdenum
Formula: C₆MoO₆
MW: 264 g/mol
Boiling Pt: 155 °C (1013 hPa)
Density: 1.96 g/cm³ (20 °C)
MDL Number: MFCD00003466
CAS Number: 13939-06-5
EINECS: 237-713-3
UN: 3466
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR