G418 disulphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
M-Clarity™ quality level = MQ200
G5013-1G G5013-250MG G5013-5G
SIALG5013-1GEA 95.3 EUR
SIALG5013-1G SIALG5013-250MG SIALG5013-5G
G418 disulphate, Sigma-Aldrich®
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2