G418 disulphate 50 mg/ml in aqueous solution, sterile filtered, Sigma-Aldrich®
Supplier: Merck
Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
M-Clarity™ quality level = MQ200
G8168-10ML G8168-100ML
SIALG8168-10MLEA 167 EUR
SIALG8168-10ML SIALG8168-100ML
G418 disulphate 50 mg/ml in aqueous solution, sterile filtered, Sigma-Aldrich®
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
MDL Number: MFCD00058314
CAS Number: 108321-42-2