Orcein

Supplier: Roth Carl

ROTH0320.1EA 0 EUR
ROTH0320.1
Orcein
Orcein
MDL Number: MFCD00062310
CAS Number: 1400-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR