Sodium hypochlorite 4-6% available chlorine in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
419550010 419550250 419550025 419550100 419552500
ACRO419550010EA 171 EUR
ACRO419550010 ACRO419550250 ACRO419550025 ACRO419550100 ACRO419552500
Sodium hypochlorite 4-6% available chlorine in aqueous solution
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Boiling Pt: 111 °C (1013 hPa)
Melting Pt: –30…–20 °C
Density: 1.083 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8 III
Merck Index: 13,08702
Order Now
Specification Test Results
Appearance (Color) Clear yellow
Appearance (Form) Solution
Titration Iodometric 4.5 to 5.5 %