Nickel(II) sulphamate 50% (w/w) in aqueous solution, Reagent Grade
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Nickel (II) sulphamidate, Nickel bis(sulphamidate), Nickel sulphamate
40501.K2 40501.8M
40501.K2EA 55.5 EUR
40501.K2 40501.8M
Nickel(II) sulphamate 50% (w/w) in aqueous solution, Reagent Grade
Nickel (II) sulphamate
CAS No.: 13770-89-3 Molecular Formula: H4N2NiO6S2 Formula Weight: 250.86(anhy) Physical Form: Liquid Appearance: Green Odour: Odourless UN No.: 3082 MDL No.: MFCD00137261
Nickel plating
Formula: H₄N₂NiO₆S₂
MW: 250.86
Storage Temperature: Ambient
MDL Number: MFCD00137261
CAS Number: 13770-89-3
EINECS: 237-396-1
UN: 3082
ADR: 9 III