(±)-Ethyl-2-methylbutyrate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(±)-2-Methylbutanoic acid ethyl ester, (±)-2-Methylbutyric acid ethyl ester, Ethyl 2-methylbutanoate
W244317-100G-K
W244317-1KG-K
W244317-20KG-K
W244317-4KG-K
SIALW244317-100G-KEA
84.1
EUR
SIALW244317-100G-K
SIALW244317-1KG-K
SIALW244317-20KG-K
SIALW244317-4KG-K
(±)-Ethyl-2-methylbutyrate, Sigma-Aldrich®
(±)-Ethyl 2-methylbutyrate
Formula:
C₇H₁₄O₂ MW: 130.19 g/mol |
MDL Number:
MFCD00012217 CAS Number: 7452-79-1 |