(±)-3,3-Dimethyl-2-butanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Pinacolyl alcohol
M-Clarity™ quality level = MQ100
136824-100G 136824-25G
SIAL136824-100GEA 465 EUR
SIAL136824-100G SIAL136824-25G
(±)-3,3-Dimethyl-2-butanol, Sigma-Aldrich®
(±)-3,3-Dimethyl-2-butanol
Formula: C₆H₁₄O
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: 5 °C
Density: 0.8347 g/cm³ (0 °C)
MDL Number: MFCD00004522
CAS Number: 464-07-3
EINECS: 207-347-9
UN: 1987
ADR: 3,III