DL-Lysine monohydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride
M-Clarity™ quality level = MQ200
L6001-100G L6001-1KG L6001-25G
SIALL6001-100GEA 147 EUR
SIALL6001-100G SIALL6001-1KG SIALL6001-25G
DL-Lysine monohydrochloride, Sigma-Aldrich®
DL-Lysine monohydrochloride
Formula: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
Melting Pt: 267
MDL Number: MFCD00064563
CAS Number: 70-53-1
EINECS: 200-739-0