Sodium chloride standardised solution 0.1 N in aqueous solution
Supplier: Thermo Fisher Scientific
Synonyms: Saline, NaCl, Sea salt, Halite
35616.AP 35616.K2
35616.APEA 40.5 EUR
35616.AP 35616.K2
Sodium chloride standardised solution 0.1 N in aqueous solution
Sodium chloride
CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Physical Form: Liquid MDL No.: MFCD00003477
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599