Tin(IV) chloride 1 mol/l dichloromethane, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Stannic chloride, Tin tetrachloride
MQ100
249955-100ML
249955-800ML
SIAL249955-100MLEA
82.4
EUR
SIAL249955-100ML
SIAL249955-800ML
Tin(IV) chloride 1 mol/l dichloromethane, Sigma-Aldrich®
Tin (IV) chloride
Formula:
Cl₄Sn MW: 260.52 g/mol Boiling Pt: 114 Melting Pt: -33 Storage Temperature: Ambient |
MDL Number:
MFCD00011242 CAS Number: 7646-78-8 EINECS: 231-588-9 Merck Index: 12,08929 |