Calcium acetate xhydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid calcium salt hydrate, Calcium di(acetate) hydrate, Calcium acetate hydrate
M-Clarity™ quality level = MQ200
C1000-100G C1000-2.5KG C1000-500G
SIALC1000-100GEA 43.3 EUR
SIALC1000-100G SIALC1000-2.5KG SIALC1000-500G
Calcium acetate xhydrate, Sigma-Aldrich®
Calcium acetate xhydrate
Formula: (H₃CCOO)₂Ca·nH₂O
MW: 176.18 g/mol
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00150019
CAS Number: 114460-21-8
EINECS: 200-540-9