Gallic acid monohydrate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate
M-Clarity™ quality level = MQ100
27645-250G-R 27645-500G-R
SIAL27645-250G-REA 140 EUR
SIAL27645-250G-R SIAL27645-500G-R
Gallic acid monohydrate, Sigma-Aldrich®
Gallic acid monohydrate
Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: ∼250 °C
Flash Pt: 250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8
EINECS: 205-749-9