Gallic acid monohydrate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate
M-Clarity™ quality level = MQ200
398225-100G
398225-500G
SIAL398225-100GEA
81.8
EUR
SIAL398225-100G
SIAL398225-500G
Gallic acid monohydrate, Sigma-Aldrich®
Gallic acid monohydrate
Formula:
C₇H₆O₅·1H₂O MW: 188.14 g/mol Melting Pt: ∼250 °C Flash Pt: 250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00149098 CAS Number: 5995-86-8 EINECS: 205-749-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...