Phosphotungstic acid hydrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
79690-100G 79690-25G 79690-500G
SIAL79690-100GEA 103 EUR
SIAL79690-100G SIAL79690-25G SIAL79690-500G
Phosphotungstic acid hydrate, Sigma-Aldrich®
Phosphotungstic acid hydrate
Formula: H₃[P(W₃O₁₀)₄]·nH₂O
Melting Pt: 107 °C
MDL Number: MFCD00011342
CAS Number: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 13,07450