D-(+)-Homoserine, Sigma-Aldrich®

Supplier: Merck

Synonyms: H-D-Hser-OH, H-D-HomoSer-OH, H-D-Homoser-OH, (R)-(+)-Homoserine, (R)-2-Amino-4-hydroxybutyric acid, D-Homoserine

M-Clarity™ quality level = MQ200

H4021-1G H4021-250MG
SIALH4021-1GEA 466 EUR
SIALH4021-1G SIALH4021-250MG
D-(+)-Homoserine, Sigma-Aldrich®
D(+)-Homoserine
Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00077786
CAS Number: 6027-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR