Potassium hexacyanoferrate(II) trihydrate, Sigma-Aldrich®
Supplier: Merck
|
|
M-Clarity™ quality level = MQ100
455989-100G
455989-25G
SIAL455989-100GEA
521
EUR
SIAL455989-100G
SIAL455989-25G
Potassium hexacyanoferrate(II) trihydrate, Sigma-Aldrich®
Potassium hexacyanoferrate (II) trihydrate
Formula:
K₄Fe(CN)₆·3H₂O Melting Pt: 70 °C Density: 1.889 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00167023 CAS Number: 14459-95-1 EINECS: 237-722-2 Merck Index: 12,07793 |