L(+)-Serine
Supplier: Molekula
Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine
24133813-1KG 24133813-100G
MOLE24133813-1KGEA 0 EUR
MOLE24133813-1KG MOLE24133813-100G
L(+)-Serine
L(+)-Serine
Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 215…225 °C
Density: 1.6 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846