L-Lysine, Sigma-Aldrich®
Supplier: Merck
62840-100G-F 62840-25G-F 62840-500G-F
SIAL62840-100G-FEA 162 EUR
SIAL62840-100G-F SIAL62840-25G-F SIAL62840-500G-F
L-Lysine, Sigma-Aldrich®
L-Lysine
Formula: C₆H₁₄N₂O₂
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 196 °C
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2
Merck Index: 13,05656