Sodium hypophosphite monohydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sodium phosphinate monohydrate
S5012-100G S5012-500G
SIALS5012-100GEA 38.3 EUR
SIALS5012-100G SIALS5012-500G
Sodium hypophosphite monohydrate, Sigma-Aldrich®
Sodium hypophosphite monohydrate
Formula: NaH₂PO₂·H₂O
Melting Pt: 90 °C
Density: 0.8 g/cm³ (20 °C)
MDL Number: MFCD06252379
CAS Number: 10039-56-2
EINECS: 231-669-9