Magnesium hydrogen orthophosphate trihydrate ≥99%
Supplier: Thermo Scientific
Synonyms: Magnesium hydrogen phosphate trihydrate, Magnesium phosphate dibasic trihydrate
A14279.0B A14279.30
A14279.0BEA 67.9 EUR
A14279.0B A14279.30
Magnesium hydrogen orthophosphate trihydrate ≥99%
Magnesium hydrogen orthophosphate trihydrate
Purity: 99% CAS No.: 7782-75-4 Molecular Formula: MgHPO4.3H2O Formula Weight: 174.34 (120.28anhy) Odour: Odourless Merck Reference: 14,5681 MDL No.: MFCD00150390
Formula: HMgO₄P·3H₂O
MW: 174.33 g/mol
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00150390
CAS Number: 7782-75-4
EINECS: 231-823-5
Merck Index: 14,5681