Indium(III) nitrate hydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Indium trinitrate hydrate

M-Clarity™ quality level = MQ100

326135-10G 326135-50G
SIAL326135-10GEA 70.6 EUR
SIAL326135-10G SIAL326135-50G
Indium(III) nitrate hydrate, Sigma-Aldrich®
Indium (III) nitrate hydrate
Formula: InN₃O₉·H₂O
MW: 318.85 g/mol
MDL Number: MFCD00149742
CAS Number: 207398-97-8
EINECS: 237-393-5
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR