Lutetium trinitrate hydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Lutetium (III) nitrate hydrate
M-Clarity™ quality level = MQ100
436429-1G 436429-5G
SIAL436429-1GEA 82 EUR
SIAL436429-1G SIAL436429-5G
Lutetium trinitrate hydrate, Sigma-Aldrich®
Lutetium trinitrate hydrate
Formula: LuN₃O₉·H₂O
MW: 379 g/mol
MDL Number: MFCD00150421
CAS Number: 100641-16-5
EINECS: 233-241-7
UN: 1477
ADR: 5.1,III