(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
295825-1G 295825-250MG 295825-5G
SIAL295825-1GEA 92.9 EUR
SIAL295825-1G SIAL295825-250MG SIAL295825-5G
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, Sigma-Aldrich®
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 241…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-56-5
Merck Index: 13,01223