Petrochemical distillation apparatus for water in crude oil
Supplier: DWK Life Sciences
201-1418EA 1060 EUR
201-1418
Petrochemical distillation apparatus for water in crude oil
Distillation Components Distillation Sets
This apparatus is used for the determination of water in crude oil by distillation.
  • Designed from ASTM D4006 specifications
  • Manufactured from borosilicate 3.3 glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Apparatus includes a 1000 ml round bottom flask with NS 24/40 joint, a distillation receiver with 0,05 graduations, a drying trap and a 400 ml Liebig condenser.