Zinc perchlorate hexahydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
401439-100G 401439-500G
SIAL401439-100GEA 38 EUR
SIAL401439-100G SIAL401439-500G
Zinc perchlorate hexahydrate, Sigma-Aldrich®
Zinc perchlorate hexahydrate
Formula: Cl₂O₈Zn·6H₂O
MW: 372.38 g/mol
Melting Pt: 106 °C
Density: 2.25 g/cm³ (20 °C)
MDL Number: MFCD00150370
CAS Number: 10025-64-6
EINECS: 237-122-0
UN: 1481
ADR: 5.1,II