L(+)-Lysine monohydrate

Supplier: Molekula

MOLEM66061013EA 0 EUR
MOLEM66061013
L(+)-Lysine monohydrate
L(+)-Lysine monohydrate
Formula: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
MW: 164,2 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00151035
CAS Number: 39665-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR