L(+)-Calcium lactate pentahydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
C8356-1KG C8356-250G C8356-500G
SIALC8356-1KGEA 568 EUR
SIALC8356-1KG SIALC8356-250G SIALC8356-500G
L(+)-Calcium lactate pentahydrate, Sigma-Aldrich®
L(+)-Calcium lactate pentahydrate
Formula: C₆H₁₀CaO₆·5H₂O
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00150344
CAS Number: 5743-47-5
EINECS: 248-953-3